Learn More
DWK Life Sciences WHEATON™ CompletePAK™ Sterile Components Stoppers
The WHEATON™ CompletePAK™ product line is a range of crimp top vials, rubber stoppers and seals that are sterilized ready-to-use and are off- the-shelf, and can be customized for any application reducing your supply chain due to our one source solution capability.
Brand: DWK Life Sciences W986005-S
Description
- Sterile
- Stopper
Certifications
USP <788>, <85> and <71>; Container Closure Integrity (CCI) certified
Specifications
Sterile | |
OmniFlex 3G Igloo | |
USP 788, 85, and 71, Container Closure Integrity (CCI) certified |
230 | |
Wheaton CompletePAK | |
13 mm |