Learn More
Corning™ Sterile Polystyrene Inoculating Loops
Constructed of flexible polystyrene for optimum retrieval of an inoculum from a culture. Gosselin™ Sterile Polystyrene Inoculating Loops provide a superior solution for transferring microscopic organisms, inoculum, or streaking.
Brand: Corning™ OS10-02
116.34 EUR valid until 2025-06-30
Use promo code "24156" to get your promotional price.
Description
- Polystyrene (PS) inoculating loops
- Spherical or needle extremity
- 1 μl or 10 μl
Specifications
10 μL | |
Polystyrene | |
Blue | |
3000/Pk. |
194 mm | |
Sterile | |
Inoculating Loop, Needle Tip | |
Tamper evident zip bag |
Promotions
-
Multiple Manufacturers
Spotlight
Big discounts are available on a range of popular products including chemicals, lab essentials, equipment, consumables, life science and safety products.Promo Code: 24156Expires: 2025-06-30
Days
28Hours
1Minutes
35Seconds
48