Learn More
Greiner Bio-One 384 Well Microplates
Designed with a rounded square profile for flexible working volume combined with reduced wicking and bubbling. Greiner Bio-One™ 384 Well Polystyrene Microplates provide four times the number of wells in a 96 well standard microplate while retaining external dimensions compatible with standard equipment and automated systems.
Brand: Greiner Bio-One 781162
This item is not returnable.
View return policy
Description
- Clear polystyrene construction for optimum visibility
- Footprint compatible with automated systems
- Can be traced to production through defined LOT number system
- Free of detectable DNase, RNase and human DNA
- Non-pyrogenic
- Manufactured under DIN ISO 9001 guidelines
- Well-to-well spacing: 4.5mm
- Square wells with rounded corners combines advantages of square well (i.e. flexible working volume of 10μ to 130μ with the advantages of a round well (reduced wicking and bubbling)
Specifications
DIN ISO 9001 | |
Standard | |
Clear | |
384 | |
Sterile | |
Flat Bottom (F-well) | |
100/Pk. |
DNase/RNase and human DNA free | |
16 x 24 | |
Polystyrene | |
10 to 130 μL | |
138 μL | |
100Pack |