Learn More
Fisherbrand™ Sterile Syringes for Single Use
Manual syringe without needle for research purposes only. Suitable for use with syringe filters
Brand:  Fisherbrand™ 14955455
 View more versions of this product
Product Code. 15839152
  • 73.70€ / Pack of 50
Edge
Description
Description
  • GAMMA sterilization
  • Sterile
  • Individually wrapped in a paper/plastic blister pack
Services
5 Years
Specifications
Specifications
29.2 mm
60 mL
Sterile
Slip Tip
Leur Slip
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Fisherbrand™ Sterile Syringes for Single Use > 60mL