Learn More
Fisherbrand™ Polystyrene Petri Dishes
GREENER_CHOICE
Clear. Fisherbrand™ Polystyrene Petri Dishes are perfect for microbial or cell culture applications.
Brand:  Fisherbrand™ FB51504
42.75 EUR valid until 2025-12-31
Use promo code "24345" to get your promotional price.
 View more versions of this product
Product Code. 12654785
  • 136.00€ / Pack of 600
In Stock
Edge
Specifications
Polystyrene
Petri Dish
1
90 mm
Non-sterile
600 Case
16.2 mm
Videos
  • Fisherbrand - Advantage aspiration system
    The Fisherbrand aspiration Advantage system is a compact and portable unit with an integrated resistant diaphragm pump, used for the safe and precise aspiration of non-flammable chemical, biological, and medical liquids.
    Video Player is loading.
    Current Time 0:00
    Duration 0:00
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:00
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
      Fisherbrand Advantage aspiration system
    SDS
    Documents Certificates
    Certificates
    A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
    2 results found
    Lot Number Certificate Type Date Catalog Number
    Lot NumberM554280 Certificate TypeCertificate of Compliance Date28/04/2025 Catalog NumberFB51504
    Lot NumberM553410 Certificate TypeCertificate of Compliance Date13/03/2025 Catalog NumberFB51504
    Promotions
    Promotions
    Product Content Correction
    Product Title
    Fisherbrand™ Polystyrene Petri Dishes > Aseptic; 1 Vent