Learn More
Sampling Systems™ SteriWare™ Disposable Liquid Funnel
Ideal for laboratories, pharmaceutical environments, cleanrooms and chemical manufactruing. Sampling Systems™ SteriWare™ Disposable Liquid Funnel comes individually bagged after being moulded and packed in a Class 100,000 Cleanroom.
Brand: Sampling Systems™ 8370H-200S
Description
- Constructed of HDPE
- Moulded and packed in a Class 100,000 Cleanroom
- Individually bagged
- Made from FDA, EC 1935/2004 and EU 10/2011 conforming material
- BSE/TSE free
- Full batch traceability
- Available pre-sterilized (SAL 10-6)
- No cross contamination
- No cleaning required
Specifications
200 mm | |
15 mm | |
Sterile | |
Filling Funnel (Liquid) |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
067647 | Certificate of Analysis | 29/01/2025 | 8370H-200S |
067646 | Certificate of Analysis | 29/01/2025 | 8370H-200S |
067647 | Certificate of Compliance | 13/01/2025 | 8370H-200S |
119326 | Certificate of Compliance | 26/11/2024 | 8370H-200S |
110686 | Certificate of Compliance | 19/06/2024 | 8370H-200S |
Product Content Correction
Product Title
Sampling Systems™ SteriWare™ Disposable Liquid Funnel > ItemDescription: Yes