Learn More
Sampling Systems™ SteriWare™ Disposable Liquid Funnel
BIOPHARMA
Ideal for laboratories, pharmaceutical environments, cleanrooms and chemical manufactruing. Sampling Systems™ SteriWare™ Disposable Liquid Funnel comes individually bagged after being moulded and packed in a Class 100,000 Cleanroom.
Brand:  Sampling Systems™ 8370H-200
 View more versions of this product
Product Code. 15889741
Description
Description
  • Constructed of HDPE
  • Moulded and packed in a Class 100,000 Cleanroom
  • Individually bagged
  • Made from FDA, EC 1935/2004 and EU 10/2011 conforming material
  • BSE/TSE free
  • Full batch traceability
  • Available pre-sterilized (SAL 10-6)
  • No cross contamination
  • No cleaning required
Specifications
Specifications
200 mm
15 mm
Non-sterile
Liquid Funnel
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number121372 Certificate TypeCertificate of Compliance Date27/02/2025 Catalog Number8370H-200
Lot Number120438 Certificate TypeCertificate of Compliance Date20/01/2025 Catalog Number8370H-200
Lot Number117343 Certificate TypeCertificate of Compliance Date26/01/2024 Catalog Number8370H-200
Lot Number071978 Certificate TypeCertificate of Compliance Date07/12/2022 Catalog Number8370H-200
Lot Number070288 Certificate TypeCertificate of Compliance Date23/11/2022 Catalog Number8370H-200
Product Title
Sampling Systems™ SteriWare™ Disposable Liquid Funnel > ItemDescription: No