Learn More
Sampling Systems™ SteriWare™ Disposable Liquid Funnel
BIOPHARMA
Ideal for laboratories, pharmaceutical environments, cleanrooms and chemical manufactruing. Sampling Systems™ SteriWare™ Disposable Liquid Funnel comes individually bagged after being moulded and packed in a Class 100,000 Cleanroom.
Brand:  Sampling Systems™ 8370H-100
 View more versions of this product
Product Code. 16259421
  • 58.85€ / Pack of 50
Only null left
Estimated Shipment: 24-07-2025
Description
  • Constructed of HDPE
  • Moulded and packed in a Class 100,000 Cleanroom
  • Individually bagged
  • Made from FDA, EC 1935/2004 and EU 10/2011 conforming material
  • BSE/TSE free
  • Full batch traceability
  • Available pre-sterilized (SAL 10-6)
  • No cross contamination
  • No cleaning required
Specifications
Specifications
100 mm
12 mm
Non-sterile
Liquid Funnel
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
4 results found
Lot Number Certificate Type Date Catalog Number
Lot Number108629 Certificate TypeCertificate of Compliance Date15/11/2024 Catalog Number8370H-100
Lot Number071773 Certificate TypeCertificate of Compliance Date20/08/2024 Catalog Number8370H-100
Lot Number108392 Certificate TypeCertificate of Compliance Date13/06/2023 Catalog Number8370H-100
Lot Number000579 Certificate TypeCertificate of Compliance Date10/02/2023 Catalog Number8370H-100
Promotions
Promotions
Product Content Correction
Product Title
Sampling Systems™ SteriWare™ Disposable Liquid Funnel > ItemDescription: No