Learn More
Sampling Systems™ SteriWare™ Disposable Powder Funnel
Constructed of chemically resistant, food grade HDPE. Sampling Systems™ SteriWare™ Disposable Powder Funnel is ideal for transferring and sampling powders.
Brand: Sampling Systems™ 8111H-200S
Description
- Moulded and packed in a Class 100,000 Cleanroom
- Individually bagged
- Made from FDA, EC 1935/2004 and EU 10/2011 conforming material
- BSE/TSE free
- Full batch traceability
- Available pre-sterilized (SAL 10-6)
- No cross contamination
- No cleaning required
Specifications
200 mm | |
HDPE | |
Sterile |
40 mm | |
10 Pk. |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
121380 | Certificate of Compliance | 27/02/2025 | 8111H-200S |
067377 | Certificate of Compliance | 13/01/2025 | 8111H-200S |
120787 | Certificate of Compliance | 16/12/2024 | 8111H-200S |
119735 | Certificate of Compliance | 18/09/2024 | 8111H-200S |
119042 | Certificate of Compliance | 11/07/2024 | 8111H-200S |
Product Content Correction
Product Title
Sampling Systems™ SteriWare™ Disposable Powder Funnel > Type: Sterile