Learn More
Lenz Laborglasintrumente Stirrer with Stirrer Bearing
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand: Lenz Laborglasintrumente 06310507
This item is not returnable.
View return policy
Description
- Shaft with blade and PTFE security screw
- Gastight closure of vacuum stirring apparatuses up to 10-3 mbar at a continuous operation speed of max.
- 800 rpm, for round bottom flasks 250-2000 mL
- Shaft made of DURAN™ tubing
Specifications
Screw cap, GL 25 | |
For round bottom flasks 250 to 2000 mL | |
Polypropylene |