Learn More
Poulten Graf™ Stopper Caps
43.05€
Specifications
Product Type Stopper Cap
Quantity 1 Ea.
View More Specs
Products 2
Product Code Brand Volume (Metric) Price Quantity & Availability  
Product Code Brand Volume (Metric) Price Quantity & Availability  
10178101
View Documents
Poulten Graf™
101 08918K
10 , 30 mL
43.05€
Pack of 10
Only null left
Estimated Shipment: 15-07-2025
Add to basket
10583622
View Documents
Poulten Graf™
101 08917K
5 mL
43.05€
Pack of 10
Only null left
Estimated Shipment: 15-07-2025
Specifications
Stopper Cap
1 Ea.
Documents
Product Certifications
Product Content Correction
Product Title
Poulten Graf™ Stopper Caps