Learn More
Thermo Scientific™ Storage Canes
GREENER_CHOICE
Store various cryostorage tubes in these storage canes, available in standard or stepped format to accommodate a variety of vial sizes and designs.
Brand:  Thermo Scientific™ 5015-0002
 View more versions of this product
Product Code. 11860491
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products »
Description
Store Thermo Scientific™ Nalgene™ or Thermo Scientifc™ Nunc™ cryostorage tubes in Thermo Scientific™ Storage Canes, available in standard or stepped format to accommodate a variety of vial sizes and designs.
  • Able to store tubes protected with Thermo Scientific™ CryoFlex™ tubing
  • Compatible with color coders that attach to the end of the cane and can be written on for quick identification and inventory management
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CryoCane; Standard; Aluminum; 300 mm
Aluminum
12/Pk., 48/Cs.
30 cm
Standard
Storage Cane
11.8125 in.