Learn More
Lenz Laborglasinstrumente™ Splash Heads Acc. to Stutzer, Straight
For placement onto distillation flasks for separation of drops. Lenz Laborglasinstrumente™ Splash Heads Acc. to Stutzer, Straight, are manufactured from durable Duran™ glass tubing.
53.32€ - 98.26€
Specifications
For Use With (Application) | For separation of drops (Direct placement onto distillation flasks) |
---|---|
Type | Straight Splash Heads |
Material | Duran Glass |
Product Code | Brand | Standard Taper Joint | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Standard Taper Joint | Price | Quantity & Availability | |||||
10588421
|
Lenz Laborglasinstrumente™
05035011 |
NS 14/23 NS 14/23 |
71.82€
Each |
Estimated Shipment: 13-11-2024
Log in to see stock. |
|||||
11953461
|
Lenz Laborglasinstrumente™
05035045 |
NS 29/32, NS 14/23 |
98.26€
Each |
Estimated Shipment: 20-11-2024
Log in to see stock. |
|||||
10231071
|
Lenz Laborglasinstrumente™
05035044 |
NS 29/32, NS 29/32 |
53.32€
Each |
Estimated Shipment: 24-10-2024
Log in to see stock. |
|||||
Description
- Straight with NS cone and socket
- Temperature and chemical resistance
- Minimal thermal expansion
Specifications
For separation of drops (Direct placement onto distillation flasks) | |
Straight Splash Heads | |
Duran Glass |