Learn More
IBA Lifesciences GmbH Strep-Tactin™ APC for Fab Streptamers™
IBA Lifesciences GmbH Strep-Tactin™ APC for Fab Streptamers™ is for reversible cell isolation delivering label-free target cells.
Brand:  IBA Lifesciences GmbH 6-5011-005
Product Code. 16431485
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products »
Description
Description
  • Low-affinity Fab-Strep molecules are reversibly multimerized on Strep-Tactin™ PE or APC forming a fluorescent Fab Streptamer™ for cell staining
  • Treatment of labeled target cells with the competing Strep-Tactin™ ligand biotin causes a disruption of the Fab Streptamer™ complex and results in spontaneous dissociation of the Fab-Streps from the target cells
  • The result is a functional and label-free target cell
Specifications
Specifications
All required reagents for cell isolation
For 5 x 107 to 1 x 108 cells
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Product Title
IBA Lifesciences GmbH Strep-Tactin™ APC for Fab Streptamers™ > Includes: All required reagents for cell isolation