Learn More
Cole-Parmer™ Stuart™ Borosilicate Glass Receiving Flask
Brand:  Cole-Parmer™ 10096-72
 View more versions of this product
Product Code. 12089121
  • 186.00€ / Each
Estimated Shipment: 04-04-2025
Specifications
Borosilicate Glass
For RE100 Rotary Evaporator
Round
Receiving Flask
1 Each
500 mL