missing translation for 'onlineSavingsMsg'
Learn More
Succinimide, 98+%, Thermo Scientific Chemicals Product Code.: 11415603

Succinimide, 98+%, Thermo Scientific Chemicals

Product Code. 11415603
1000 g
Click to view available options
Quantity:
250 g
1000 g
5000 g
This item is not returnable. View return policy

Product Code. 11415603

Brand: Thermo Scientific Alfa Aesar A13503.0B

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 123-56-8 | C4H5NO2 | 99.089 g/mol

Succinimide is used in organic syntheses and in industrial silver plating processes. It is also used as anticonvulsant drugs such as ethosuximide, phensuximide and methsuximide. Further, it is used to form covalent bonds between proteins, peptides and plastics. In addition to this, it is used as an important raw material for the synthesis of heterocyclic compounds through N-acyliminium cyclizations.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Succinimide is used in organic syntheses and in industrial silver plating processes. It is also used as anticonvulsant drugs such as ethosuximide, phensuximide and methsuximide. Further, it is used to form covalent bonds between proteins, peptides and plastics. In addition to this, it is used as an important raw material for the synthesis of heterocyclic compounds through N-acyliminium cyclizations.

Solubility
Soluble in water and ethanol. Insoluble in ether and chloroform.

Notes
Incompatible with strong oxidizing agents, acids, bases and strong reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 123-56-8
Molecular Formula C4H5NO2
Molecular Weight (g/mol) 99.089
MDL Number MFCD00005495
InChI Key KZNICNPSHKQLFF-UHFFFAOYSA-N
Synonym succinimide, 2,5-pyrrolidinedione, butanimide, succinic acid imide, 2,5-dioxopyrrolidine, 2,5-diketopyrrolidine, succinic imide, succinimide-sauba, 3,4-dihydropyrrole-2,5-dione, dihydro-3-pyrroline-2,5-dione
PubChem CID 11439
ChEBI CHEBI:9307
IUPAC Name pyrrolidine-2,5-dione
SMILES C1CC(=O)NC1=O

Specifications

Melting Point 121°C to 126°C
Density 1.41
pH 4 to 6
Boiling Point 287°C to 288°C
Flash Point 201°C (393°F)
Odor Odorless
Quantity 1000 g
Beilstein 108440
Merck Index 14,8871
Solubility Information Soluble in water and ethanol. Insoluble in ether and chloroform.
Formula Weight 99.09
Percent Purity ≥98%
Chemical Name or Material Succinimide
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10254689 Certificate TypeCertificate of Analysis Date17/05/2025 Catalog NumberA13503.30,A13503.0I,A13503.0B
Lot NumberX03K007 Certificate TypeCertificate of Analysis Date22/10/2024 Catalog NumberA13503.30,A13503.0I,A13503.0B
Lot NumberS08J054 Certificate TypeCertificate of Analysis Date24/10/2023 Catalog NumberA13503.30,A13503.0I,A13503.0B
Lot NumberM12J024 Certificate TypeCertificate of Analysis Date18/01/2023 Catalog NumberA13503.30,A13503.0I,A13503.0B
Lot NumberT19I021 Certificate TypeCertificate of Analysis Date29/07/2022 Catalog NumberA13503.30,A13503.0I,A13503.0B

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Succinimide, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.