Learn More
Sucrose (Brix) Standard, 40%, Reagecon™
BIOPHARMA
Brand:  Reagecon BS40S
Quantity 15 mL
Product Code. 13474036
  • 71.33€ / 15mL
Edge
Specifications
40%
15 mL
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberRIBS4025D1 Certificate TypeCertificate of Analysis Date17/06/2025 Catalog NumberBS40S
Lot NumberRIBS4025B1 Certificate TypeCertificate of Analysis Date27/04/2025 Catalog NumberBS40S
Lot NumberRIBS4021E1 Certificate TypeCertificate of Analysis Date28/03/2025 Catalog NumberBS40S
Lot NumberRIBS4024L1 Certificate TypeCertificate of Analysis Date14/12/2024 Catalog NumberBS40S
Lot NumberRIBS4024J1 Certificate TypeCertificate of Analysis Date23/10/2024 Catalog NumberBS40S
Promotions
Promotions
Product Content Correction
Product Title
Sucrose (Brix) Standard, 40%, Reagecon™ > Quantity: 15mL