Learn More
Tocris Bioscience™ Sulfidefluor 7 AM
H2S fluorescent probe
Brand:  Tocris Bioscience™ 4943/1
Product Code. 18749748
  • 436.00€ / 1mg
Estimated Shipment: 30-01-2025
Description
Sulfidefluor 7 AM is a reported to be a hydrogen sulphide (H2S) fluorescent probe. Contains an acetoxymethyl ester, which enhances cell permeability and is hydrolyzed inside the cell to trap the probe. Used to visualize VEGF-induced H2S production in human umbilical vein endothelial cells (HUVECS). Excitation wavelength ∽ 495 nm; emission maximum ∽ 520 nm.
Specifications
Specifications
1 mg
Sulfidefluor 7 AM
685.55
Sulfidefluor 7 AM