missing translation for 'onlineSavingsMsg'
Learn More

Chemetrics™ Sulfite (free) - Titrets™ Titration Cells

Achieve accurate titrations. Chemetrics™ Sulfite (free) - Titrets™ Titration Cells are ideal for performing accurate titrations. The titrets are dependable, fast, and easy with no need to count drops.

Brand:  Chemetrics™ K9610


Product Code. 11741014

  • 41.00€ / Pack of 30
Estimated Shipment: 10-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Dependable, sealed single-use titrants
  • Fast, easy procedure -- no need to count drops
  • No burettes or other equipment necessary
  • Perform a titration ANYWHERE
Specifications

Specifications

30 ampoules with valve assemblies, Neutralizer Solution, titrettor, 25mL sample cup, and instructions
30
10 to 100ppm
Iodometric
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • X30 TITRETS SULFITE 10-100 PPM

Signal Word
  • Danger

Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 1 A
  • Skin corrosion/irritation Category 1 B
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 1

Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H319-Causes serious eye irritation.
  • H372-Causes damage to organs.
  • H411-Toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P270-Do not eat, drink or smoke when using this product.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P321-Specific treatment (see on this label).
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P363-Wash contaminated clothing before reuse.
  • P391-Collect spillage.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : sodium hydroxide, EDTA disodium salt, potassium iodide, potassium iodate, sulphamic acid, phosphoric

SDS
Documents

Documents

Certificates
Special Offers

Special Offers