Learn More
Witeg™ Sulfonation Flasks
An excellent vessel for use in pharmaceutical applications.
Brand: Witeg™ 0 850 031
This item is not returnable.
View return policy
Description
- Available in either 100 or 200 mL capacities
- With 4 side necks
- Center neck socket ST 29/32
- Side necks socket ST 19/26
Specifications
100 mL | |
Pharmaceutical | |
Sulfonation flask |
Glass | |
4 side necks, 1 center | |
CN ST 29/32, SN ST 19/26 |