Learn More
Sulfur, AAS standard solution, Specpure™ S 1000μg/mL, Thermo Scientific Chemicals Product Code.: 11318237
Sulfur, AAS standard solution, Specpure™ S 1000μg/mL, Thermo Scientific Chemicals
Product Code. 11318237
100 mL
Click to view available options
Quantity:
100 mL
500 mL
Product Code. 11318237
Brand: Thermo Scientific Alfa Aesar 022958.AE
Request Bulk or Custom Format
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
MDL Number MFCD00085316
Specifications
Quantity 100 mL
Physical Form Liquid
Grade Specpure™
Chemical Name or Material Sulfur AAS standard solution, S 1000μg/mL
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number1550384 Certificate TypeCertificate of Analysis Date10/04/2025 Catalog Number022958.AP,022958.AE
Lot Number1538951 Certificate TypeCertificate of Analysis Date11/03/2025 Catalog Number022958.AP,022958.AE
Lot Number1514419 Certificate TypeCertificate of Analysis Date20/12/2024 Catalog Number022958.AP,022958.AE
Lot Number1503403 Certificate TypeCertificate of Analysis Date26/11/2024 Catalog Number022958.AP,022958.AE
Lot Number1496723 Certificate TypeCertificate of Analysis Date10/10/2024 Catalog Number022958.AP,022958.AE
Product Title
Sulfur, AAS standard solution, Specpure™ S 1000μg/mL, Thermo Scientific Chemicals >