Learn More
Bollé Safety SUPERBLAST Industrial Spare Lens, Clear Platinum
Product Code. 16812330
For use with SUPERBLAST Clear safety goggle
PLATINUM coating : anti-scratch & anti-fog
TRUSTED_SUSTAINABILITY
Specifications
Lens Type Platinum
Thickness (Metric) 2 mm
Product Type Spare lens
Disposable Reusable
Lens Material Polycarbonate