Learn More
3M™ Swab-Sampler
Convenient and quick collection device for environmental sampling and testing over small surface areas.
Brand:  3M™ 7000053817
Encompass
7333.61 EUR valid until 2024-11-30
Use promo code "21864" to get your promotional price.
 View more versions of this product
Product Code. 17203734
  • 146.13€ / Pack of 100
Estimated Shipment: 13-02-2025
Description
  • Ready to use for pathogen testing of environmental samples.
  • Swab reaches into corners and hard-to-reach areas for increased accuracy.
  • Convenient design helps improve sampling consistency from tech-to-tech and plant-to-plant.
  • Bottles have ample headspace for sample addition and vortexing.
  • Self-contained swab and vial for increased productivity and efficiency.
Specifications
Specifications
10 mL Letheen Broth
100
Swab