missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Syntide 2, 96%

For research use

62.95€ - 299.49€

Specifications

For Use With (Application) Enzyme Assay Substrate
Formulation Lyophilized powder
Storage Requirements -30° to -10°C
Regulatory Status RUO
Protein Length Pro-Leu-Ala-Arg-Thr-Leu-Ser-Val-Ala-gly-Leu-Pro-gly-Lys-Lys
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15485149
View Documents
Thermo Scientific Chemicals
J62843.M^
1mg
62.95€
1mg
Please to purchase this item. Need a web account? Register with us today!
15475149
View Documents
Thermo Scientific Chemicals
J62843.#0
5mg
299.49€
5mg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Selective substrate for calmodulin-dependent protein kinase II

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Enzyme Assay Substrate
-30° to -10°C
Pro-Leu-Ala-Arg-Thr-Leu-Ser-Val-Ala-gly-Leu-Pro-gly-Lys-Lys
Native
96%
Lyophilized powder
RUO
Syntide 2, 96%
Lyophilized
Videos
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific Chemicals Syntide 2, 96%

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.