Learn More
Terumo™ Syringe Ste Tuberculin
Syringe Ste Tuberculin is easy to use and can be used in a variety of applications.
Brand:  Terumo™ 8MDSS01S2516E
Product Code. 17122714
  • 25.65€ / Pack of 100
Edge
Description
  • Easy to use
TRUSTED_SUSTAINABILITY
Specifications
Specifications
25 G
Syringe Ste Tuberculin
1 mL
15 mm
100 Pack
Product Title
Terumo™ Syringe Ste Tuberculin > Volume: 1 mL