Learn More
Tocris Bioscience™ T 112
Estrogen-related receptor γ agonist; promotes maturation of hiPSC-derived cardiomyocytes
Brand: Tocris Bioscience™ 7690/50
587.46 EUR valid until 2024-12-16
Use promo code "21630" to get your promotional price.
This item is not returnable.
View return policy
Description
T 112 is an estrogen-related receptor gamma (ERRγ) agonist. Promotes maturation of hiPSC-derived cardiomyocytes. T 112 accelerates metabolic and sarcomere maturation in cardiomyocytes, as well as maturation of contractile and electrophysiological properties and ultrastructure organization, when used alone or in combination with other small molecules.Specifications
2-[2-(4-Bromophenyl)-1H-benzimidazol-6-yl]-2,3-dihydro-5-(4-morpholinyl)-1H-isoindol-1-one | |
50 mg | |
C25H21BrN4O2 |
2376876-87-6 | |
Estrogen and Related Receptor Agonists |