Learn More
Brady™ Tamper-evident Inspection Date Labels
Inspection date labels can be used wherever a frequent and regular inspection of machinery and equipment is required.
Brand:  Brady™ 834143
 View more versions of this product
Product Code. 15898801
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products »
Explore more special offers
Description
Description
  • For indication of inspection dates, such as on fire extinguishers
  • Simply mark label with hole punch and apply wherever you need it
  • Use the yearly color system to easily identify when your equipment was last inspected
  • Tamper-evident labels show evidence of unauthorized removal
Specifications
Specifications
Not Autoclavable
White on Purple
Polyolefin
2023
104
Round
0.076 mm
Permanent Rubber
Satin
Non-sterile
Inventory and Inspection Labeling, General Identification, Safety Identification
104 Labels
-70°C to +80°C
10 mm