Learn More
Applied Biosystems™ TAMRA™ Dye Calibration Plate, 384-well
The 384-Well Dye Calibration Plate (TAMRA™) is used to maintain your real-time PCR instrument with 384-well block
Brand: Applied Biosystems™ 4443619
This item is not returnable.
View return policy
Description
- The 384-Well Dye Calibration Plate (TAMRA™) is used to maintain your real-time PCR instrument with 384-well block.
- Dye calibration should be run after lamp change.
- Please see your instrument user guide for full instructions on use.
Specifications
384-well Dye Calibration Plate | |
TAMRA™, TAMRA™ | |
Dye Calibration Plate | |
Dry Ice | |
Spectral Calibration |
Store at –20°C ± 5°C. |
|
ViiA™ 7 Dx System, QuantStudio™ Dx | |
Real Time PCR (qPCR) | |
1 x 384 well plate | |
qPCR |