Learn More
Tocris Bioscience™ Targapremir 210
Inhibits formation of miR-210
Brand:  Tocris Bioscience™ 6586/5
Product Code. 18716566
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products »
Explore more special offers
Description
Description
Targapremir 210 inhibits formation of mature miR-210 (IC50 = 200 nM) by binding to Dicer binding site of the miR-210 hairpin precursor (Kd = 200 nM) and inhibits Dicer processing of the miRNA. Decreases levels of miR-210 and HIF-1α mRNA, and induces apoptosis in MDA-MB-231 cells cultured under hypoxic conditions. Reduces tumor expression of HIF-1α mRNA and miR-210, and tumor growth in a triple negative breast cancer mouse model.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
N-(3-Azidopropyl)-4-[3-[6-(4-methyl-1-piperazinyl)[2,6'-bi-1H-benzimidazol]-2'-yl]phenoxy]butanamide
5 mg
98%
1049722-30-6
C32H36N10O2
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Tocris Bioscience™ Targapremir 210 > 5 mg