missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tartrate Resistant Acid Phosphatase Monoclonal Antibody (7E6A11), Invitrogen™
Mouse Monoclonal Antibody
Brand: Thermo Scientific MA548357
This item is not returnable.
View return policy
Description
Tartrate-resistant acid phosphatase (TRAP) is a basic iron-binding protein with high activity towards phosphoproteins, ATP and 4-nitrophenyl phosphatase. It is detected in human alveolar macrophages, osteoclasts, spleen and liver. Its expression is increased in spleen and monocytes of patients with Gaucher's disease; spleenocytes and circulating leukocytes of patients with hairy cell leukemia; spleen of patients with Hodgkin's lymphoma and in the sera of patients undergoing active bone turnover.
Specifications
Tartrate Resistant Acid Phosphatase | |
Monoclonal | |
1 mg/mL | |
PBS with 0.05% sodium azide | |
P13686 | |
ACP5 | |
Purified recombinant fragment of human ACP5 (AA: 221-325) expressed in E. Coli. | |
100 μg | |
Primary | |
Human | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
7E6A11 | |
Unconjugated | |
ACP5 | |
acid phosphatase 5, tartrate resistant; acid phosphatase type V; Acp5; HPAP; human purple acid phosphatase; T5ap; Tartrate-resistant acid ATPase; tartrate-resistant acid phosphatase 5a; tartrate-resistant acid phosphatase 5b; tartrate-resistant acid phosphatase type 5; TRACP; TRACP5a; TRACP5b; TRAP; TR-AP; TRAPase; TrATPase; TTRRAP; Type 5 acid phosphatase | |
Mouse | |
Protein G | |
RUO | |
54 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Tartrate Resistant Acid Phosphatase Monoclonal Antibody (7E6A11), Invitrogen™ > 100 μg; Unconjugated
Spot an opportunity for improvement?Share a Content Correction