missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TC-I 15, Tocris Bioscience™
Click to view available options
Quantity:
10 mg
50 mg
Description
TC-I 15 is a potent α2β1 integrin inhibitor (IC50 values for the inhibition of human platelet adhesion to type I collagen are 12 and 715 nM for platelets under static conditions and under flow, respectively). TC-I 15 displays selectivity for α2β1 over αvβ3, α5β1, α6β1 and αIIbβ3 at concentrations exceeding 1000 nM. TC-I 15 reduces collagen IV production in mesangial cells. Active in vivo; TC-I 15 prevents ferric chloride-induced clot formation in mice.
Specifications
Specifications
Quantity | 10 mg |
Formula Weight | 520.62 |
Percent Purity | >98% |
Chemical Name or Material | TC-I 15 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction