Learn More
Technical Buffer for pH DKD Certified pH 4.01, Mettler Toledo™
Shop All Mettler Toledo Products
Click to view available options
Quantity:
250 mL
Description
Greener Choice Claims
- Environmental benefits include:
- Certifications and Seals of Approval
- Renewable Energy
Specifications
Specifications
pH | 4.01 |
Quantity | 250 mL |
Packaging | 1Each |
Promotions
-
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13
-
Multiple Manufacturers
Quality Analysis and Quality Control Testing
Save up to 59% on some of our most popular productsPromo Code: 24140Expires: 2025-08-24
New Offer Save Up to 59%
Product Content Correction
Product Title
Technical Buffer for pH DKD Certified pH 4.01, Mettler Toledo™ >