Learn More
Aalborg™ Model F PTFE/PFA Flowmeter
Aalborg™ Model F PTFE/PFA Flowmeter is excellent for high-purity applications or use with corrosive liquids.
Brand:  Aalborg™ F6C-F02-01-TF
 View more versions of this product
Product Code. 11717729
  • 621.00€ / Each
Only null left
Estimated Shipment: 27-08-2025
Description
  • Chemically inert wetted components
  • Removable protective shield
  • Individually leak tested
  • Vertical in-line
  • Valve at outlet
Specifications
Specifications
Flowmeter
6.7 bar
Ultra-pure or corrosive liquids
121°C
250 mL/min.
±5%
1/4 in. NPT (M)
Linear and direct
Teflon
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Aalborg™ Model F PTFE/PFA Flowmeter > Flow Rate Range: 250mL/min.