Learn More
Buerkle™ Telescopic Rod
Versatile sampling system with exchangeable tools for a wide range of applications.
36.10€ - 125.00€
Specifications
Product Type | Telescopic Rod |
---|---|
Material | Aluminum |
Autoclavable | Not Autoclavable |
For Use With (Application) | Industry and Water Treatment, Ideal for Sampling From Baths, Tanks, Shafts, Open Water Courses, Used with Telescope Sampler |
For Use With (Equipment) | Bürkle Angled beaker, pendulum beaker, bottle holder, stainless steel beaker and dip net |
Product Code | Brand | Length (Metric) | Includes | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Includes | Price | Quantity & Availability | |||||
10396242
|
Buerkle™
5355-0120 |
65 to 120 cm (Adjustable) | 2 Rods |
36.10€
Each |
In Stock
Log in to see stock. ![]() |
|||||
10530053
|
Buerkle™
5355-0250 |
125 to 250 cm (Adjustable) | 2 Rods |
46.15€
Each |
In Stock
Log in to see stock. ![]() |
|||||
15376297
|
Buerkle™
5355-0180 |
70 to 180 cm | 4 Rods |
61.05€
Each |
In Stock
Log in to see stock. |
|||||
10376292
|
Buerkle™
5355-0280 |
95 to 280 cm (Adjustable) | 4 Rods |
72.10€
Each |
In Stock
Log in to see stock. |
|||||
10040822
|
Buerkle™
5355-0300 |
115 to 300 cm (Adjustable) | 3 Rods |
72.10€
Each |
In Stock
Log in to see stock. ![]() |
|||||
10182412
|
Buerkle™
5355-0450 |
165 to 450 cm (Adjustable) | 3 Rods |
101.00€
Each |
In Stock
Log in to see stock. ![]() |
|||||
10244992
|
Buerkle™
5355-0600 |
175 to 600 cm (Adjustable) | 4 Rods |
125.00€
Each |
In Stock
Log in to see stock. |
|||||
Description
- Different tools can be attached to the telescopic rod
- The tools (angular beaker, pendulum beaker, bottle holder, stainless steel beaker and dip net) are attached to the telescopic rod by a practical snap-in joint
- Ideal for sampling from baths, tanks, shafts, open water courses etc
- Sampling depths down to 6 m can be reached with the adjustable telescopic aluminum rod
- The telescopic rods cannot be rotated radially, which means even better handling
- The individual rods have been provided with an end stop, this means that one rod cannot accidentally slip out of another rod
- Telescopic rod 5355-0280, extremely short model – yet extendable up to 280 cm. Fits into every car boot!
- Springs made of stainless steel
Specifications
Telescopic Rod | |
Not Autoclavable | |
Bürkle Angled beaker, pendulum beaker, bottle holder, stainless steel beaker and dip net |
Aluminum | |
Industry and Water Treatment, Ideal for Sampling From Baths, Tanks, Shafts, Open Water Courses, Used with Telescope Sampler |
Product Title
Buerkle™ Telescopic Rod