Learn More
BINDER™ Temperature Calibration Certificate for CB Incubators
This Temperature Calibration Certificate is for use with BINDER™ CB Incubators with Hot Air Sterilization and Heat Sterilizable CO2 Sensor. A variety of temperature measurement options are available.
440.00€ - 1735.00€
Specifications
For Use With (Equipment) | BINDER Chambers CB Incubators with Hot Air Sterilization and Heat Sterilizable CO2 Sensor |
---|---|
Duration | 1 Year |
Calibration Type | Temperature |
Product Type | Calibration Certificate |
Product Line | BINDER™ CB |
Product Code | Brand | Calibration Points | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Calibration Points | Price | Quantity & Availability | |||||
16424089
|
BINDER™
8012-1132 |
Central Measuring Point at Specified Temperature |
440.00€
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16474279
|
BINDER™
8012-1550 |
9 Measuring Points (at Specified Temperature) |
1050.00€
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16414299
|
BINDER™
8012-1571 |
15 to 18 Measuring Points (at Specified Temperature) |
1325.00€
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16484309
|
BINDER™
8012-1592 |
27 Measuring Points (at Specified Temperature) |
1735.00€
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Temperature calibration with certificate with a variety of measurement options
- For use with BINDER™ CB Incubators with Hot Air Sterilization and Heat Sterilizable CO2 Sensor
Specifications
BINDER Chambers CB Incubators with Hot Air Sterilization and Heat Sterilizable CO2 Sensor | |
Temperature | |
BINDER™ CB | |
Temperature Calibration |
1 Year | |
Calibration Certificate | |
Operational Qualification |
Product Title
BINDER™ Temperature Calibration Certificate for CB Incubators