Learn More
Greiner Bio-One Terasaki Plates for Microscopic Examinations
Suitable for serological determination of Human Leukocyte Antigens. Greiner Bio-One Terasaki Plates are used for microscopic examinations.
Brand: Greiner Bio-One 659190
This item is not returnable.
View return policy
Description
- Individual well volume of 10μL
- Flat well bottom
- Improved cell adhesion through physical surface treatment
- Space saver design with recessed lid
- Available in high throughput folding carton packaging or inner wrappe
Specifications
6 x 10 | |
Closure Included | |
Flat | |
Clear | |
Rectangular | |
Tissue Culture | |
60 | |
1200 | |
1200/Cs. | |
57.8 mm |
Clear | |
Polystyrene | |
10 L (Well) | |
Polystyrene | |
Non-sterile | |
Microtest Plate | |
Lid | |
Inner packs of 150 | |
83.2 mm | |
8 mm |