Learn More
Thermo Scientific Chemicals Tergitol(TM) 15-S-9 Product Code.: 16358237
compliance-icons compliance-icons
Thermo Scientific Chemicals Tergitol(TM) 15-S-9
Product Code. 16358237
1 L
Click to view available options
Quantity:
250 mL
1 L
2.5 L
10 L
Product Code. 16358237
Brand: Thermo Scientific Chemicals 464250010
Request Bulk or Custom Format
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 68131-40-8
Molecular Formula [C2H4O]n C13H27O
MDL Number MFCD00132411
SMILES CCCCCCCC(CCCCC)OCCO-*
Specifications
Density 1.0100 g/mL
Quantity 1 L
Flash Point 193°C
Specific Gravity 1.01
Solubility Information Solubility aq. soln.: soluble.
Chemical Name or Material Tergitol* 15-S-9
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
4 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0466805 Certificate TypeCertificate of Analysis Date16/10/2024 Catalog Number464250025,464252500,464250010,464250100
Lot NumberA0406829 Certificate TypeCertificate of Analysis Date19/02/2024 Catalog Number464250025,464252500,464250010,464250100
Lot NumberA0456800 Certificate TypeCertificate of Analysis Date09/10/2023 Catalog Number464250025,464252500,464250010,464250100
Lot NumberA0406829 Certificate TypeCertificate of Analysis Date20/06/2022 Catalog Number
compliance-icons compliance-icons
Product Identifier
  • Tergitol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Tergitol
Product Title
Thermo Scientific Chemicals Tergitol(TM) 15-S-9 >