missing translation for 'onlineSavingsMsg'
Learn More
tert-Butyl 4-[5-(hydroxymethyl)-4-methyl-1,3-thiazol-2-yl]piperidine-1-carboxylate, ≥97%, Thermo Scientific™ Product Code.: 10321043

tert-Butyl 4-[5-(hydroxymethyl)-4-methyl-1,3-thiazol-2-yl]piperidine-1-carboxylate, ≥97%, Thermo Scientific™

Product Code. 10321043
1 g, Amber Glass Bottle, 1g
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 10321043

Brand: Thermo Scientific Maybridge CC31109DA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 857283-66-0
Molecular Formula C15H24N2O3S
Molecular Weight (g/mol) 312.428
MDL Number MFCD07368658
InChI Key YNLQKGMVENCCLE-UHFFFAOYSA-N
Synonym tert-butyl 4-5-hydroxymethyl-4-methyl-1,3-thiazol-2-yl tetrahydro-1 2h-pyridinecarboxylate, tert-butyl 4-5-hydroxymethyl-4-methyl-1,3-thiazol-2-yl piperidine-1-carboxylate, 1-piperidinecarboxylicacid, 4-5-hydroxymethyl-4-methyl-2-thiazolyl-, 1,1-dimethylethyl ester, 1-tert-butoxycarbonyl-4-5-hydroxymethyl-4-methyl-1,3-thiazol-2-yl piperidine, 4-5-hydroxymethyl-4-methyl-thiazol-2-yl-piperidine-1-carboxylic acid tert-butyl ester, 4-5-hydroxymethyl-4-methyl-1,3-thiazol-2-yl piperidine, n1-boc protected, tert-butyl 4-5-hydroxymethyl-4-methylthiazol-2-yl piperidine-1-carboxylate
PubChem CID 7164575
IUPAC Name tert-butyl 4-[5-(hydroxymethyl)-4-methyl-1,3-thiazol-2-yl]piperidine-1-carboxylate
SMILES CC1=C(SC(=N1)C2CCN(CC2)C(=O)OC(C)(C)C)CO

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 312.43
Percent Purity 95%
Chemical Name or Material Tert-butyl 4-[5-(hydroxymethyl)-4-methyl-1, 3-thiazol-2-yl]tetrahydro-1(2H)-pyridinecarboxylate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
tert-Butyl 4-[5-(hydroxymethyl)-4-methyl-1,3-thiazol-2-yl]piperidine-1-carboxylate, ≥97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.