Learn More
Julabo™ Test Tube Rack
Test tube rack made of high grade plastic or stainless steel
Brand: Julabo™ 9970321
This item is not returnable.
View return policy
Specifications
Test tube racks for water baths | |
42 | |
Up to +150 °C |
Stainless Steel | |
75 x 12/13 mm dia. |
Product Suggestions
Customers who viewed this item also viewed.
Product Content Correction
Product Title
Julabo™ Test Tube Rack > No. of Positions: 42