Learn More
Texwipe™ Cleanroom Polypropylene Buckets
Brand:  Texwipe™ TX7062
 View more versions of this product
Product Code. 17905241
  • 97.40€ / Each
Estimated Shipment: 14-03-2025
Specifications
22 L
Green
Flat Mops
24 cm
45 cm
24 cm
Polypropylene
18 x 9.5 x 9.5 in. (45 x 24 x 24 cm)
9.5 in.
18 in.
9.5 in.