missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TFF1/pS2 Antibody (TFF1/7768), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-21010AF750
This item is not returnable.
View return policy
Description
TFF1/pS2 Monoclonal antibody specifically detects TFF1/pS2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
TFF1/pS2 | |
Monoclonal | |
Alexa Fluor 750 | |
BCEIbreast cancer, estrogen-inducible sequence expressed in, Breast cancer estrogen-inducible protein, breast cancer estrogen-inducible sequence, D21S21, gastrointestinal trefoil protein pS2, hP1.A, HPS2, pNR-2, Polypeptide P1.A, Protein pS2, pS2, trefoil factor 1, trefoil factor, BCE1, human pS2 induced by estrogen from human breast cancercell line M10HP1.A | |
Recombinant fragment (around aa1-84) of human TFF1/pS2 protein (exact sequence is proprietary) | |
0.1 mL | |
Breast Cancer, Cancer | |
7031 | |
Store at 4°C in the dark. | |
IgG |
Immunohistochemistry (Paraffin) | |
TFF1/7768 | |
50 mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
TFF1/pS2 Antibody (TFF1/7768), Alexa Fluor™ 750, Novus Biologicals™ > 0.1 mL, Alexa Fluor 750
Spot an opportunity for improvement?Share a Content Correction