Learn More
TGF beta 1/2/3 Antibody (1D11), mFluor Violet 450 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals FAB1835MFV450
Product Code. 30060701
  • 654.00€ / 0.10mL
Estimated Shipment: 11-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
TGF beta 1/2/3 Monoclonal antibody specifically detects TGF beta 1/2/3 in Broad Species Reactivity samples. It is validated for Western Blot, Binding Inhibition, ELISA Standard (Matched Pair), Neutralization, ELISA (Matched Antibody Pair), ELISA (Matched Antibody Pair)
Specifications
Specifications
TGF beta 1/2/3
Monoclonal
mFluor Violet 450 SE
50mM Sodium Borate
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
7040
Store at 4C in the dark.
IgG1
Western Blot, Binding Inhibition Assay, ELISA Standard (Matched Pair), Neutralization, ELISA Capture (Matched Antibody Pair)
1D11
Western Blot, Binding Inhibition, ELISA Standard (Matched Pair), Neutralization, ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair)
CED, DPD1, LAP, TGFB, TGFB1, TGFbeta, transforming growth factor, beta 1
Bovine bone-derived TGF-beta 1 and TGF-beta 2
0.1 mL
Primary
Broad Species Reactivity