missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TGF beta 1/2/3 Antibody (1D11), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB1835MFV610
This item is not returnable.
View return policy
Description
TGF beta 1/2/3 Monoclonal antibody specifically detects TGF beta 1/2/3 in Broad Species Reactivity samples. It is validated for Western Blot, Binding Inhibition, ELISA Standard (Matched Pair), Neutralization, ELISA (Matched Antibody Pair), ELISA (Matched Antibody Pair)Specifications
TGF beta 1/2/3 | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
7040 | |
Store at 4C in the dark. | |
IgG1 |
Western Blot, Binding Inhibition Assay, ELISA Standard (Matched Pair), Neutralization, ELISA Capture (Matched Antibody Pair) | |
1D11 | |
Western Blot, Binding Inhibition, ELISA Standard (Matched Pair), Neutralization, ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair) | |
CED, DPD1, LAP, TGFB, TGFB1, TGFbeta, transforming growth factor, beta 1 | |
Bovine bone-derived TGF-beta 1 and TGF-beta 2 | |
0.1 mL | |
Primary | |
Broad Species Reactivity | |
Purified |