Learn More
Applied Biosystems™ Thermal Tape (for the 310 Genetic Analyzer)
This is a roll of 0.5 ft. yellow lab marking thermal tape for affixing the capillary to the 310 Genetic Analyzer heat plate.
Brand: Applied Biosystems™ 310021
This item is not returnable.
View return policy
Description
- For Research Use Only
- Not for use in diagnostics procedures
Specifications
Store at room temperature. | |
310 Genetic Analyzer | |
Tape | |
Room Temperature |