Learn More
Thermo Scientific™ Evolution™ One, OnePlus and Pro Spectrophotometer Solid Sample Holders
Designed to hold solid samples for transmittance measurements
1075.00€ - 1695.00€
Specifications
For Use With (Equipment) | Thermo Scientific™ Evolution™ One, OnePlus and Pro UV-visible spectrophotometers |
---|
Product Code | Brand | Holds | Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Holds | Type | Price | Quantity & Availability | ||||
10061104
|
Thermo Scientific™
222-217400 |
Round or square optical samples, glass or Plastic samples, lenses from sunglasses or goggles | Universal Sample Holder |
1075.00€
Each |
Estimated Shipment: 20-02-2025
Log in to see stock. |
||||
11801892
|
Thermo Scientific™
222-217200 |
Standard 2 x 3 in. sample holders | Solid Sample Holder |
1695.00€
Each |
Estimated Shipment: 20-02-2025
Log in to see stock. |
||||
Description
Designed to hold solid samples for transmittance measurements
Solid Sample Holder
- Provides easy mounting of solid samples like filters, plastic, glass and optics
- Accommodates standard 2 x 3 in. sample holders
- Holds round or square optical samples; glass or plastic samples; lenses from sunglasses or goggles
- Requires one Solid Sample Holder (14-387-503)
Specifications
Thermo Scientific™ Evolution™ One, OnePlus and Pro UV-visible spectrophotometers |