Learn More
Thermo Scientific™ Nalgene™ Replacement Closures for Economy Wash Bottles
Click to view available options
Neck Finish:
24-400
28-400
Description
- Closure and stem molded in one piece for leakproof service
- Tip can be cut back to provide increased flow
- Tight fit of draw tube and closure prevents separation when squeezing bottle
- Leakproofâ€
Greener Choice Claims
- Environmental benefits include:
- Renewable Energy
Specifications
Specifications
Material | Polypropylene |
Neck Finish | 24-400 |
For Use With (Equipment) | Wash bottle |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
1 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
7452972010 | Certificate of Compliance | 17/04/2025 | 712401-0002 |
Product Content Correction
Product Title
Thermo Scientific™ Nalgene™ Replacement Closures for Economy Wash Bottles >