Learn More
Eppendorf ThermoMixer™ C
Shop All Eppendorf Products
Click to view available options
Type:
Dry Bath/Block
Description
Reliable prevention of condensate formation in small vessels can be achieved with the heated lid Eppendorf ThermoTop (optional). All common vessels and plate formats from 5μL to 50mL by different Eppendorf SmartBlocks for standard applications.
- Excellent 2DMix-Control technology results in excellent sample mixing
- Anti-spill technology prevents lid wetting and cross-contamination
- Increased mixing frequency of up to 3,000 rpm for effective sample mixing
- Excellent temperature management for maximum temperature accuracy, resulting in safe handling
- Compatible with Eppendorf ThermoTop (condens.protect™) for condensation-free sample handling (for Eppendorf SmartBlocks up to 2.0mL)
- Predefined program keys and clearly arranged menu guides enable simple and intuitive operations
- Freely programmable program keys for individual optimization
- Eppendorf PhysioCare Concept provides ergonomic design and operation
Specifications
Specifications
Temperature Range (Metric) | Ambient -15° to 100°C |
Type | Dry Bath/Block |
Display | Temperature Settings: 1°C/100°C |
Speed | 300 to 3000rpm |
Timer | 15 sec. to 99:30 hr., continuous |
Length (Metric) | 30.4 cm |
Width (Metric) | 20.6 cm |
Height (Metric) | 13.1 cm |
Depth (Metric) | 30.4 cm |
Voltage | 220/240 V |
Show More |
Special Offers
- Multiple Manufacturers
-
Multiple Manufacturers
Back to the Lab Summer Semester
Save up to 94% on Teaching Lab EssentialsPromo Code: 25721Expires: 2025-05-01
New Offer Save Now
Product Content Correction
Product Title
Eppendorf ThermoMixer™ C >