Learn More
Thiophosgene, 85%, AcroSeal™ Product Code.: 10783055
Thiophosgene, 85%, AcroSeal™
Product Code. 10783055 Shop All Thermo Scientific Chemicals Products
100 mL, AcroSeal™ Glass bottle, 100mL
Click to view available options
Quantity:
100 mL
Packaging:
AcroSeal™ Glass bottle
Unit Size:
100mL
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 10783055
Brand: Thermo Scientific Acros 426441000
Request Bulk or Custom Format
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
CAS: 463-71-8 | CCl2S | 114.98 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifications
Boiling Point 73.0°C
CAS 463-71-8
Color White through Yellow
Physical Form Solid
Quantity 100 mL
Assay Percent Range 80% min. (GC)
Molecular Formula CCl2S
Linear Formula ClCSCl
MDL Number MFCD00004918
Flash Point 62°C
Synonym thiophosgene, carbonothioic dichloride, carbon chlorosulfide, thiocarbonyl chloride, thiocarbonic dichloride, dichlorothiocarbonyl, thiofosgen, phosgene, thio, thiokarbonylchlorid, carbonyl chloride, thio
Solubility Information Solubility in water: slow decomposition in cold, fast in hot water. Other solubilities: soluble in ether, ccl4, decomposes in alcohol
InChI Key ZWZVWGITAAIFPS-UHFFFAOYSA-N
SMILES C(=S)(Cl)Cl
IUPAC Name thiocarbonyl dichloride
Molecular Weight (g/mol) 114.98
PubChem CID 10040
ChEBI CHEBI:29366
Formula Weight 114.98
Percent Purity 85%
Packaging AcroSeal™ Glass bottle
Refractive Index 1.5442
Specific Gravity 1.508
Density 1.5080g/mL
Chemical Name or Material Thiophosgene
Show More Show Less
compliance-icons
Product Identifier
  • Thiophosgene
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H330-Fatal if inhaled.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Product Title
Thiophosgene, 85%, AcroSeal™ >