Learn More
Novus Biologicals™ TLR (Intracellular) Screening Antibody Pack
For Research applications
Brand:  Novus Biologicals™ NBP2-25085
Product Code. 18121292
  • 491.00€ / Pack of 6
Estimated Shipment: 19-12-2024
to see stock.
Explore more special offers
Add to basket
Includes:
  • 1.NBP2-24729: TLR9 Antibody (26C593.2)
  • 2.NBP2-24875: TLR3 Antibody (40C1285.6)
  • 3.NBP2-24906: TLR7 Antibody
  • 4.NBP2-24917: TLR8 Antibody (44C143)
Specifications
TLR (Intracellular) Screening
Unconjugated
TLR3: 25 ug: WB, Flow, IP, IHC: H: Purified TLR7: 25 ug: WB, Flow, IHC, IP, IF/ICC: H, M: Purified TLR8: 25 ug: WB, Flow, IHC: H, M: Purified TLR9: 25 ug: WB, Flow, IHC, IF/ICC, CM: H, M, R, RM: Purified WB:Western Blot IHC:Immunohistochemistry IP:Immunoprecipitation CM:Confocal Microscopy FA-Neut Functional Assay Neutralization IF/ICC:Immunofluorescence/Immunocytochemistry D:Dog H:Human M:Mouse Mk:Monkey R:Rat RM:Rhesus Monkey
1.NBP2-24729: TLR9 Antibody (26C593.2)
2.NBP2-24875: TLR3 Antibody (40C1285.6)
3.NBP2-24906: TLR7 Antibody
4.NBP2-24917: TLR8 Antibody (44C143)
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, KnockDown
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Dot Blot, Flow (Intracellular)
4 Vials
Human, Mouse