Learn More
Cole-Parmer™ TNP-5 Torque Meter
Quality control and quality assurance instrument for food, beverage, cosmetics and other torque applications
Brand: Cole-Parmer™ 59891-07
This item is not returnable.
View return policy
Description
- Sample size from 10-190mm diameter
- Measures opening and closing torque values
- Store 1000 data points
- USB Interface
- Rechargeable NiMH batteries or AC adapter
Specifications
TNP-5 digital torque meter | |
For measuring opening and closing torque values | |
Rechargeable NiMH batteries or AC adapter | |
320 x 227 x 126 mm |
Product Content Correction
Product Title
Cole-Parmer™ TNP-5 Torque Meter > Dimensions (L x W x H): 320 x 227 x 126mm